Gene Transcripts

Gene Symbol
Nipsnap3a
Entrez Gene ID
73398
Gene Name
nipsnap homolog 3A (C. elegans)
Gene Aliases
1700054F22Rik, AI429120
Chromosome Location
Chr. 4: 52989284 - 53000854 on Build GRCm38
UniGene ID
Mm.252738
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028529.1 NP_082805.1 2 364
GenBank mRNA AK006782.1 - 2 340

View Full Product Details