Gene Transcripts

Gene Symbol
NDUFA3
Entrez Gene ID
4696
Gene Name
NADH:ubiquinone oxidoreductase subunit A3
Gene Aliases
B9, CI-B9
Chromosome Location
Chr. 19: 54102338 - 54106974 on Build GRCh38
UniGene ID
Hs.198269
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004542.3 NP_004533.1 3 154
XM_017026833.1 XP_016882322.1 4 571
GenBank mRNA AF044955.1 - 3 146
AF070653.1 - 3 155
AK289489.1 - 3 152
BC022369.1 - 3 151 4737656
BC061644.1 - 3 154 5748578
CR542143.1 - 2 127

View Full Product Details