Gene Transcripts

Gene Symbol
Abhd14b
Entrez Gene ID
76491
Gene Name
abhydrolase domain containing 14b
Gene Aliases
1810013B01Rik
Chromosome Location
Chr. 9: 106447737 - 106452923 on Build GRCm38
UniGene ID
Mm.335427
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029631.3 NP_083907.3 3, 4 550
GenBank mRNA AK007469.1 - 3, 4 505
BC019410.1 - 3, 4 529 4191819

View Full Product Details