Gene Transcripts

Gene Symbol
Slc9a8
Entrez Gene ID
77031
Gene Name
solute carrier family 9 (sodium/hydrogen exchanger), member 8
Gene Aliases
1200006P13Rik, 6430709P13Rik, AI182282, NHE-8, NHE8
Chromosome Location
Chr. 2: 167420056 - 167477000 on Build GRCm38
UniGene ID
Mm.278924
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AB089793.1 - 6, 7 563
AF482993.1 - 5, 6 527
AK037377.1 - 6 1444
AK049410.1 - 6, 7 559
AK051791.1 - 5, 6 478
AK143281.1 - 6, 7 532
AK173069.1 - 6, 7 899
BC030879.1 - 6, 7 562 4241988
BC058947.1 - 6, 7 533 5702781

View Full Product Details