Gene Transcripts

Gene Symbol
Filip1l
Entrez Gene ID
78749
Gene Name
filamin A interacting protein 1-like
Gene Aliases
4631422O05Rik, Doc1
Chromosome Location
Chr. 16: 57353277 - 57572804 on Build GRCm38
UniGene ID
Mm.328360
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001040397.4 NP_001035487.2 3, 4 610
GenBank mRNA AK146705.1 - 3, 4 611
EU704258.1 - 3, 4 457

View Full Product Details