Gene Transcripts

Gene Symbol
Celf3
Entrez Gene ID
78784
Gene Name
CUGBP, Elav-like family member 3
Gene Aliases
4930415M08Rik, BRUNOL1, CAGH4, ERDA4, Tnrc4
Chromosome Location
Chr. 3: 94478285 - 94492199 on Build GRCm38
UniGene ID
Mm.44292
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001289613.1 NP_001276542.1 8 1450
NM_001289616.1 NP_001276545.1 9 932
NM_001289620.1 NP_001276549.1 9 932
NM_172434.3 NP_766022.1 8 1447
GenBank mRNA AK019578.1 - 5 1358
AK030259.1 - 7 758
AK044069.1 - 8 880
AK044506.1 - 9 896
AK087536.1 - 9 894
AY165052.1 - 8 1165
BC031740.1 - 5 611
BC057553.1 - 8 1505 6401356

View Full Product Details