Gene Transcripts

Gene Symbol
Wwox
Entrez Gene ID
80707
Gene Name
WW domain-containing oxidoreductase
Gene Aliases
5330426P09Rik, 9030416C10Rik, WOX1
Chromosome Location
Chr. 8: 114439630 - 115352712 on Build GRCm38
UniGene ID
Mm.440420
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019573.3 NP_062519.2 6 695
GenBank mRNA AF187014.1 - 6 646
AK018507.1 - 6 625
AK019911.1 - 6 653
AK050541.1 - 6 647
AK078528.1 - 6 648
BC014716.1 - 6 651 4241066
EF410109.1 - 1 313

View Full Product Details