Gene Transcripts
- Gene Symbol
- Selk
- Entrez Gene ID
- 80795
- Gene Name
- selenoprotein K
- Gene Aliases
- 1110001C03Rik, AA673253, HSPC030, Hsp30
- Chromosome Location
- Chr. 14: 29968380 - 29975074 on Build GRCm38
- UniGene ID
- Mm.104491
- Species
- Mus musculus
- Gene Symbol
- BC052688
- Entrez Gene ID
- 432812
- Gene Name
- cDNA sequence BC052688
- Gene Aliases
- Chromosome Location
- Chr. 13: 61771580 - 61787282 on Build GRCm38
- UniGene ID
- Mm.317358
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC066226.1 |
- |
|
2498 |
|
View Full Product Details