Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_053145.2 | NP_444375.2 | 1 | 2498 | |
GenBank mRNA | AF326313.1 | - | 1 | 2474 | |
AK045399.1 | - | 1 | 2499 | ||
AK045527.1 | - | 1 | 2471 | ||
AK046137.1 | - | 1 | 2491 | ||
AY013782.1 | - | 1 | 2339 | ||
BC031895.1 | - | 1 | 2433 | 5364116 |