Gene Transcripts

Gene Symbol
Ssx2ip
Entrez Gene ID
99167
Gene Name
synovial sarcoma, X breakpoint 2 interacting protein
Gene Aliases
AU014939, AU042321, Adip
Chromosome Location
Chr. 3: 146404617 - 146440144 on Build GRCm38
UniGene ID
Mm.200783
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001253768.1 NP_001240697.1 9, 10 1470
NM_001253769.1 NP_001240698.1 8, 9 1366
NM_001253770.1 NP_001240699.1 8, 9 1215
NM_138744.3 NP_620083.1 8, 9 1215
GenBank mRNA AF532969.1 - 7, 8 1010
AK031356.1 - 9, 10 1194
AK043865.1 - 8, 9 1319
AK049080.1 - 8, 9 1367
AK077112.1 - 9, 10 1255
AK173064.1 - 8, 9 1202
BC021749.1 - 8, 9 1170 4165430
BC031527.1 - 8, 9 1178 4008624

View Full Product Details