Gene Transcripts

Gene Symbol
ACVR2B
Entrez Gene ID
93
Gene Name
activin A receptor type 2B
Gene Aliases
ACTRIIB, ActR-IIB, HTX4
Chromosome Location
Chr. 3: 38454299 - 38493142 on Build GRCh38
UniGene ID
Hs.174273
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106.3 NP_001097.2 7 872
XM_005265583.3 XP_005265640.1 7 3334
XM_017007514.1 XP_016863003.1 7 3313
XM_017007515.1 XP_016863004.1 7 958
XM_017007516.1 XP_016863005.1 7 4282
GenBank mRNA AB209449.1 - 6 4637
BC096243.1 - 7 852 40005760
BC096244.1 - 7 852 40005761
BC096245.1 - 6 934 40005763
BC099642.4 - 7 852 40005762
X77533.1 - 7 852

View Full Product Details