Gene Transcripts

Gene Symbol
Rab31
Entrez Gene ID
106572
Gene Name
RAB31, member RAS oncogene family
Gene Aliases
1700093E07Rik, AI415285, Rab22B
Chromosome Location
Chr. 17: 65651726 - 65772752 on Build GRCm38
UniGene ID
Mm.29274
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133685.2 NP_598446.2 2, 3 337
GenBank mRNA AB232617.1 - 2, 3 129
AK051270.1 - 2, 3 257
AK156677.1 - 2, 3 338
AK159095.1 - 2, 3 226
BC013063.1 - 2, 3 266 3497938

View Full Product Details