Gene Transcripts

Gene Symbol
Prl2c5
Entrez Gene ID
107849
Gene Name
prolactin family 2, subfamily c, member 5
Gene Aliases
MRP-4, Mrpplf4, PLF-4
Chromosome Location
Chr. 13: 13182703 - 13191930 on Build GRCm38
UniGene ID
Mm.392548
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_181852.1 NP_862900.1 2 79
GenBank mRNA AF128884.1 - 2 79
AK145824.1 - 3 274
BC132514.1 - 2 111 40130791
BC132516.1 - 2 111 40130793

View Full Product Details