Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_181406.3 | NP_852071.2 | 11 | 973 | |
GenBank mRNA | AK075721.1 | - | 11 | 954 | |
AK156182.1 | - | 11 | 951 | ||
AK166986.1 | - | 11 | 1489 | ||
BC024878.1 | - | 11 | 953 | 3989666 |