Gene Transcripts
- Gene Symbol
- Asic5
- Entrez Gene ID
- 63866
- Gene Name
- acid-sensing (proton-gated) ion channel family member 5
- Gene Aliases
- Accn5, Blinac, Inac
- Chromosome Location
- Chr. 2: 180935392 - 180964365 on Build Rnor_6.0
- UniGene ID
- Rn.44631
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_022227.1 |
NP_071563.1
|
3 |
455 |
|
GenBank mRNA |
Y19034.1 |
- |
3 |
455 |
|
View Full Product Details