Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_014726.2 | NP_055541.1 | 3 | 1220 | |
XM_005257859.4 | XP_005257916.1 | 4 | 519 | ||
XM_005257860.4 | XP_005257917.1 | 4 | 1344 | ||
XM_005257861.4 | XP_005257918.1 | 4 | 824 | ||
XM_005257862.3 | XP_005257919.1 | 4 | 444 | ||
XM_017025431.1 | XP_016880920.1 | 3 | 1728 | ||
XM_017025432.1 | XP_016880921.1 | 3 | 1724 | ||
GenBank mRNA | AB018318.1 | - | 3 | 1220 | |
BC111418.1 | - | 3 | 799 | ||
BC152407.1 | - | 3 | 1220 | 8862510 | |
BC167150.1 | - | 3 | 1220 | 8862522 |