Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_019564.3 | NP_062510.2 | 2 | 554 | |
GenBank mRNA | AF172994.1 | - | 2 | 530 | |
AF179369.1 | - | 2 | 525 | ||
AK090320.1 | - | 2 | 269 | ||
AK128916.1 | - | 2 | 527 | ||
AK132270.1 | - | 2 | 527 | ||
AK167667.1 | - | 2 | 120 | ||
BC013516.1 | - | 2 | 524 | 4235912 |