The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001337388.1 | NP_001327521.1 | 9 - 10 | 12541 | 70 |
NM_001337389.1 | NP_001327522.1 | 9 - 10 | 12469 | 70 | |
NM_001337390.1 | NP_001327520.1 | 9 - 10 | 12412 | 70 | |
NM_111093.2 | NP_186875.2 | 9 - 10 | 12413 | 70 |