Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LAC14
Entrez Gene ID
830795
Gene Name
laccase 14
Gene Aliases
AT5G09360, T5E8.160, T5E8_160, laccase 14
Chromosome Location
Chr.5: 2906426 - 2908658 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_120972.1 NP_196498.1 4 - 5 650 66

View Full Product Details