Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TES
Entrez Gene ID
823396
Gene Name
ATP binding microtubule motor family protein
Gene Aliases
ARABIDOPSIS NPK1-ACTIVATING KINESIN 2, AT3G43210, ATNACK2, NACK2, NPK1-ACTIVATING KINESIN 2, TETRASPORE
Chromosome Location
Chr.3: 15190786 - 15196215 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001339092.1 NP_001319676.1 12 - 13 1496 90
NM_001339093.1 NP_001327900.1 12 - 13 1670 90
NM_001339094.1 NP_001327899.1 11 - 12 1677 90
NM_001339095.1 NP_001327901.1 10 - 11 1722 90
NM_114189.3 NP_189907.2 11 - 12 1515 90
GenBank mRNA AB088121.1 - 10 - 11 1227 90

View Full Product Details