Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TES
Entrez Gene ID
823396
Gene Name
ATP binding microtubule motor family protein
Gene Aliases
ARABIDOPSIS NPK1-ACTIVATING KINESIN 2, AT3G43210, ATNACK2, NACK2, NPK1-ACTIVATING KINESIN 2, TETRASPORE
Chromosome Location
Chr.3: 15190786 - 15196215 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001339092.1 NP_001319676.1 13 - 14 1578 94
NM_001339093.1 NP_001327900.1 13 - 14 1752 94
NM_001339094.1 NP_001327899.1 12 - 13 1759 94
NM_001339095.1 NP_001327901.1 11 - 12 1804 94
NM_114189.3 NP_189907.2 12 - 13 1597 94
GenBank mRNA AB088121.1 - 11 - 12 1309 94

View Full Product Details