Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PP2A-3
Entrez Gene ID
818850
Gene Name
protein phosphatase 2A-3
Gene Aliases
AT2G42500, MHK10.22, PP2A-4, protein phosphatase 2A-3, protein phosphatase 2A-4
Chromosome Location
Chr.2: 17697626 - 17701417 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001202803.2 NP_001189732.1 4 - 5 502 107
NM_001336978.1 NP_001324021.1 4 - 5 502 107
NM_001336979.1 NP_001324022.1 4 - 5 502 107
NM_129811.4 NP_565974.1 4 - 5 493 107
NM_201943.2 NP_973672.1 2 - 3 352 107
GenBank mRNA BT010166.1 - 4 - 5 302 107
M96841.1 - 4 - 5 490 107

View Full Product Details