Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GIN1
Entrez Gene ID
504972
Gene Name
gypsy retrotransposon integrase 1
Gene Aliases
-
Chromosome Location
Chr.7: 102112337 - 102142130 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024473.2 NP_001019644.2 3 - 4 310 130
GenBank mRNA BC114679.1 - 3 - 4 310 130
BT021780.1 - 3 - 4 375 130

View Full Product Details