Product Details
- SNP ID
-
rs116266545
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:44148077 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTCCAGCACGCCCAGTGCTGACAGC[C/T]GTCCATCATGCATAGGAGCAGCTCC
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
C17orf53
PubMed Links
Gene Details
- Gene
- C17orf53
- Gene Name
- chromosome 17 open reading frame 53
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001171251.2 |
365 |
Missense Mutation |
CGT,TGT |
R92C |
NP_001164722.1 |
NM_001321310.1 |
365 |
Missense Mutation |
CGT,TGT |
R92C |
NP_001308239.1 |
NM_001321311.1 |
365 |
Missense Mutation |
CGT,TGT |
R92C |
NP_001308240.1 |
NM_024032.4 |
365 |
Missense Mutation |
CGT,TGT |
R92C |
NP_076937.2 |
XM_011525187.1 |
365 |
Missense Mutation |
CGT,TGT |
R92C |
XP_011523489.1 |
XM_011525188.1 |
365 |
Missense Mutation |
CGT,TGT |
R92C |
XP_011523490.1 |
XM_011525189.1 |
365 |
Missense Mutation |
CGT,TGT |
R92C |
XP_011523491.1 |
XM_011525190.1 |
365 |
Missense Mutation |
CGT,TGT |
R92C |
XP_011523492.1 |
XM_011525191.2 |
365 |
Missense Mutation |
CGT,TGT |
R92C |
XP_011523493.1 |
XM_011525192.1 |
365 |
Missense Mutation |
CGT,TGT |
R92C |
XP_011523494.1 |
XM_011525193.1 |
365 |
Missense Mutation |
CGT,TGT |
R92C |
XP_011523495.1 |
XM_011525194.1 |
365 |
Missense Mutation |
CGT,TGT |
R92C |
XP_011523496.1 |
XM_011525195.1 |
365 |
Missense Mutation |
CGT,TGT |
R92C |
XP_011523497.1 |
XM_011525197.2 |
365 |
Intron |
|
|
XP_011523499.1 |
XM_017025031.1 |
365 |
UTR 5 |
|
|
XP_016880520.1 |
XM_017025032.1 |
365 |
UTR 5 |
|
|
XP_016880521.1 |
View Full Product Details