Product Details

SNP ID
hCV161809915
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:6872665 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCCCTTGAATAGCCCCTCTTACC[C/T]CGGGCCCCATCAGTGCTCTGGGCCA
Phenotype
MIM: 611658 MIM: 190450 MIM: 601447
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC105369632 PubMed Links

Gene Details

Gene
LOC105369632
Gene Name
uncharacterized LOC105369632
There are no transcripts associated with this gene.

Gene
SPSB2
Gene Name
splA/ryanodine receptor domain and SOCS box containing 2
There are no transcripts associated with this gene.

Gene
TPI1
Gene Name
triosephosphate isomerase 1
There are no transcripts associated with this gene.

Gene
USP5
Gene Name
ubiquitin specific peptidase 5
There are no transcripts associated with this gene.

View Full Product Details