Product Details
- SNP ID
-
rs141356502
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.18:31318212 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AACATCTACTGAGAAATTATTTTAA[C/T]CAGACACCAGCTGAGTGGGAGAAAG
- Phenotype
-
MIM: 125670
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
DSG1
PubMed Links
Gene Details
- Gene
- DSG1
- Gene Name
- desmoglein 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001942.3 |
124 |
UTR 5 |
|
|
NP_001933.2 |
View Full Product Details