Product Details

SNP ID
rs111319230
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:32202047 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCTACCAGCCACAGCGGCTCCTTG[A/G]CCCTAGGACTTCCTCATCTGCCATC
Phenotype
MIM: 610162 MIM: 608676
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCDC28B PubMed Links

Gene Details

Gene
CCDC28B
Gene Name
coiled-coil domain containing 28B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301011.1 1216 Missense Mutation ACC,GCC T38A NP_001287940.1
NM_024296.4 1216 Missense Mutation ACC,GCC T38A NP_077272.2
XM_011542113.2 1216 Missense Mutation ACC,GCC T38A XP_011540415.1
XM_017002306.1 1216 Missense Mutation ACC,GCC T38A XP_016857795.1
XM_017002307.1 1216 Missense Mutation ACC,GCC T38A XP_016857796.1
XM_017002308.1 1216 Missense Mutation ACC,GCC T38A XP_016857797.1
XM_017002309.1 1216 Missense Mutation ACC,GCC T38A XP_016857798.1
Gene
DCDC2B
Gene Name
doublecortin domain containing 2B
There are no transcripts associated with this gene.

Gene
IQCC
Gene Name
IQ motif containing C
There are no transcripts associated with this gene.

Gene
TMEM234
Gene Name
transmembrane protein 234
There are no transcripts associated with this gene.

Gene
TXLNA
Gene Name
taxilin alpha
There are no transcripts associated with this gene.

View Full Product Details