Product Details

SNP ID
rs121434643
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:147908094 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACGGCCGCAGAGTTCGTGTGGGGG[A/G]ATGAGCAATCCGACTTCGTGTGCAA
Phenotype
MIM: 600897
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GJA8 PubMed Links

Gene Details

Gene
GJA8
Gene Name
gap junction protein alpha 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005267.4 939 Missense Mutation AAT,GAT N47D NP_005258.2
XM_011509416.1 939 Missense Mutation AAT,GAT N47D XP_011507718.1
XM_011509417.1 939 Missense Mutation AAT,GAT N47D XP_011507719.1

View Full Product Details