Product Details

SNP ID
rs138003529
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:67619052 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACTGGAGGTCCCGGCAAATCTTCA[C/T]CTCCATCAATGTCTGGAGCAAATGA
Phenotype
MIM: 611361
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
UBA6 PubMed Links

Gene Details

Gene
UBA6
Gene Name
ubiquitin like modifier activating enzyme 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018227.5 3065 Missense Mutation GAT,GGT D1035G NP_060697.4
XM_017008359.1 3065 UTR 3 XP_016863848.1

View Full Product Details