Product Details

SNP ID
rs144687989
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:35042441 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGATGGTGGCAGCTTGGGAGGTTG[C/T]GACAGCTTCATCCCTGGTGCTCTGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C9orf131 PubMed Links

Gene Details

Gene
C9orf131
Gene Name
chromosome 9 open reading frame 131
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040410.2 93 Intron NP_001035500.1
NM_001040411.2 93 Intron NP_001035501.1
NM_001040412.2 93 Nonsense Mutation CGA,TGA R15* NP_001035502.1
NM_001287391.1 93 Intron NP_001274320.1
NM_203299.3 93 Nonsense Mutation CGA,TGA R63* NP_976044.2

View Full Product Details