Product Details

SNP ID
rs200551069
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:12651207 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGATAGCGTGAGAATTAAAAAGGAC[C/T]CTGAACTTGTGGTGACCGACCTGCG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AADACL4 PubMed Links

Gene Details

Gene
AADACL4
Gene Name
arylacetamide deacetylase like 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001013630.1 253 Missense Mutation CCT,TCT P85S NP_001013652.1
XM_017001152.1 253 Missense Mutation CCT,TCT P18S XP_016856641.1
XM_017001153.1 253 Intron XP_016856642.1

View Full Product Details