Product Details

SNP ID
rs3751955
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.17:79780274 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CTTTCCCAGCCAGGGTCAATCTGGA[A/C]GGTTTGTTCTCTGGTAGCAAAAGGT
Phenotype
MIM: 602770
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CBX2 PubMed Links

Gene Details

Gene
CBX2
Gene Name
chromobox 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005189.2 Intron NP_005180.1
NM_032647.3 Intron NP_116036.1
XM_011525382.1 Intron XP_011523684.1
XM_011525383.2 Intron XP_011523685.1

View Full Product Details