Product Details

SNP ID
rs1991707
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:114156395 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCGTCTCGGTGTGTTCCGGTGTCTC[A/G]GTGTCTGCAGGTCCCAAGCCTGTAC
Phenotype
MIM: 608461
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
COL27A1 PubMed Links

Gene Details

Gene
COL27A1
Gene Name
collagen type XXVII alpha 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032888.3 Intron NP_116277.2
XM_006717308.2 Intron XP_006717371.1
XM_006717310.3 Intron XP_006717373.1
XM_011519138.2 Intron XP_011517440.1
XM_011519140.1 Intron XP_011517442.1
XM_011519142.2 Intron XP_011517444.1
XM_011519143.2 Intron XP_011517445.1
XM_011519144.2 Intron XP_011517446.1
XM_011519145.2 Intron XP_011517447.1
XM_017015239.1 Intron XP_016870728.1

View Full Product Details