Product Details

SNP ID
rs2854447
Assay Type
Validated
NCBI dbSNP Submissions
28
Location
Chr.1:225824576 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
GCCCAGCAGGCTGCCATCAGGCCTC[T/C]GAGCTCCTTCGCCAGGCGGCTTTAT
Phenotype
MIM: 132810
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
EPHX1 PubMed Links

Gene Details

Gene
EPHX1
Gene Name
epoxide hydrolase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000120.3 Intron NP_000111.1
NM_001136018.3 Intron NP_001129490.1
NM_001291163.1 Intron NP_001278092.1

View Full Product Details