Product Details
- SNP ID
-
rs2780270
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
6
- Location
-
Chr.1:112467653 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TACCTAATTTTCTCCTTTTCTTCTC[A/G]ATACATAATACTTTACATATTTATG
- Phenotype
-
MIM: 615100
MIM: 601968
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CTTNBP2NL
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs142980721] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CTTNBP2NL
- Gene Name
- CTTNBP2 N-terminal like
There are no transcripts associated with this gene.
- Gene
- MIR4256
- Gene Name
- microRNA 4256
There are no transcripts associated with this gene.
- Gene
- WNT2B
- Gene Name
- Wnt family member 2B
View Full Product Details