Product Details

SNP ID
rs9907722
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:79780008 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CGGATTGGGCCAGACTTGTTTCTGT[A/G]ATTCTAGATGTCATTTCTATTTTTA
Phenotype
MIM: 602770
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CBX2 PubMed Links

Gene Details

Gene
CBX2
Gene Name
chromobox 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005189.2 Intron NP_005180.1
NM_032647.3 Intron NP_116036.1
XM_011525382.1 Intron XP_011523684.1
XM_011525383.2 Intron XP_011523685.1

View Full Product Details