Product Details

SNP ID
rs10489761
Assay Type
Functionally tested
NCBI dbSNP Submissions
39
Location
Chr.1:35604145 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTCCTGCTCTTATCTTCCTACTCTC[C/T]TACTAAACCACGAGGTCAAGTTCAT
Phenotype
MIM: 602175 MIM: 614428
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PSMB2 PubMed Links

Gene Details

Gene
PSMB2
Gene Name
proteasome subunit beta 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199779.1 Intron NP_001186708.1
NM_001199780.1 Intron NP_001186709.1
NM_002794.4 Intron NP_002785.1
Gene
TFAP2E
Gene Name
transcription factor AP-2 epsilon
There are no transcripts associated with this gene.

View Full Product Details