Product Details

SNP ID
rs2337235
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.18:23998882 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAAAATTCCTTTTATCCTGCTGCTC[A/G]GGAACCTGTCATTATGGGGAGAGTA
Phenotype
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
TTC39C PubMed Links

Gene Details

Gene
TTC39C
Gene Name
tetratricopeptide repeat domain 39C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135993.1 Intron NP_001129465.1
NM_001243425.1 Intron NP_001230354.1
NM_001292030.1 Intron NP_001278959.1
NM_153211.3 Intron NP_694943.2
XM_005258198.4 Intron XP_005258255.1
XM_006722392.3 Intron XP_006722455.1
XM_011525814.2 Intron XP_011524116.1
XM_017025550.1 Intron XP_016881039.1
Gene
TTC39C-AS1
Gene Name
TTC39C antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details