Product Details

SNP ID
rs28735406
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.4:67616687 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTCCTTCCACACCTAATAACTGCT[C/G]TATCTCCATGATTGCAAACTTGAAA
Phenotype
MIM: 604298 MIM: 611361
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
STAP1 PubMed Links

Gene Details

Gene
STAP1
Gene Name
signal transducing adaptor family member 1
There are no transcripts associated with this gene.

Gene
UBA6
Gene Name
ubiquitin like modifier activating enzyme 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018227.5 5521 UTR 3 NP_060697.4
XM_017008359.1 5521 Intron XP_016863848.1

View Full Product Details