Product Details

SNP ID
rs10753410
Assay Type
Functionally tested
NCBI dbSNP Submissions
53
Location
Chr.1:225820400 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACCACGCCTGTCCCCAAGTATATTT[C/T]CTTAGCTCACTCTTTTCAGCATGTA
Phenotype
MIM: 132810
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EPHX1 PubMed Links

Gene Details

Gene
EPHX1
Gene Name
epoxide hydrolase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000120.3 Intron NP_000111.1
NM_001136018.3 Intron NP_001129490.1
NM_001291163.1 Intron NP_001278092.1

View Full Product Details