Product Details

SNP ID
rs1479112
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:13375520 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCACCTTCCTGCTTGGCTGGAATAC[A/C]AACCTTGCAGTAACCTTGTGAGCAA
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LINC01559 PubMed Links

Gene Details

Gene
LINC01559
Gene Name
long intergenic non-protein coding RNA 1559
There are no transcripts associated with this gene.

View Full Product Details