Product Details

SNP ID
rs1804237
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:35603313 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAAATGTTATCCAGGTCATGGATG[A/C]CATTTTTGTCAATGATTCGAACACT
Phenotype
MIM: 602175 MIM: 614428
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PSMB2 PubMed Links

Gene Details

Gene
PSMB2
Gene Name
proteasome subunit beta 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199779.1 604 Missense Mutation GGC,GTC G162V NP_001186708.1
NM_001199780.1 604 Missense Mutation GGC,GTC G70V NP_001186709.1
NM_002794.4 604 Missense Mutation GGC,GTC G187V NP_002785.1
Gene
TFAP2E
Gene Name
transcription factor AP-2 epsilon
There are no transcripts associated with this gene.

View Full Product Details