Product Details

SNP ID
rs1048447
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:67617754 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATGGTCTGAGCAGGAGAGTAGAAGG[A/G]AAGGAGCTAGAGGGAAATTACTTCC
Phenotype
MIM: 611361
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
UBA6 PubMed Links

Gene Details

Gene
UBA6
Gene Name
ubiquitin like modifier activating enzyme 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018227.5 4454 UTR 3 NP_060697.4
XM_017008359.1 4454 Intron XP_016863848.1

View Full Product Details