Product Details

SNP ID
rs260497
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:58280847 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTCCTTGCCTCATGGCCCTTTGCC[C/T]GTCTTCAAAGCCTGCATTGTAGCAT
Phenotype
MIM: 194532
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC105372482 PubMed Links

Gene Details

Gene
LOC105372482
Gene Name
uncharacterized LOC105372482
There are no transcripts associated with this gene.

Gene
ZNF544
Gene Name
zinc finger protein 544
There are no transcripts associated with this gene.

Gene
ZNF8
Gene Name
zinc finger protein 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021089.2 Intron NP_066575.2

View Full Product Details