Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nasp-2
Entrez Gene ID
179810
Gene Name
SHNi-TPR domain-containing protein
Gene Aliases
CELE_C50B6.2
Chromosome Location
Chr.V: 13310186 - 13312267 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_073897.7 NP_506298.1 3 - 4 1059 78

View Full Product Details