Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
gei-3
Entrez Gene ID
184597
Gene Name
protein-coding
Gene Aliases
CELE_T22H6.6
Chromosome Location
Chr.X: 12795201 - 12813639 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083303.5 NP_001076772.1 2 - 3 150 57

View Full Product Details