Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srx-80
Entrez Gene ID
3564918
Gene Name
7TM_GPCR_Srx domain-containing protein
Gene Aliases
CELE_F43A11.4
Chromosome Location
Chr.V: 11593727 - 11594971 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001028744.1 NP_001023915.1 6 - 7 816 77

View Full Product Details