Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FZD6
Entrez Gene ID
403610
Gene Name
frizzled class receptor 6
Gene Aliases
-
Chromosome Location
Chr.13: 4732541 - 4771294 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003065.1 NP_001003065.1 - 1975 67
XM_005627800.3 XP_005627857.1 - 2270 67
XM_005627801.3 XP_005627858.1 - 2235 67
XM_005627802.3 XP_005627859.1 - 2242 67
GenBank mRNA AY052750.1 - - 1975 67

View Full Product Details