Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NDUFAB1
Entrez Gene ID
102170842
Gene Name
NADH:ubiquinone oxidoreductase subunit AB1
Gene Aliases
-
Chromosome Location
Chr.25: 21191805 - 21202933 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018040962.1 XP_017896451.1 2 - 3 343 93
GenBank mRNA JO375465.2 - 3 - 4 350 93
JO412308.2 - 2 - 3 349 93

View Full Product Details