Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ROPN1L
Entrez Gene ID
102168771
Gene Name
rhophilin associated tail protein 1 like
Gene Aliases
-
Chromosome Location
Chr.20: 62810615 - 62829132 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005694858.3 XP_005694915.1 5 - 6 738 83
GenBank mRNA JO444597.2 - 1 - 2 182 83

View Full Product Details